missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SEC22L3 Partial ORF (NP_116752, 3 a.a. - 68 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00009117-Q01.25ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16129975

  • 528.00€ / 25µg
Spedizione stimata: 26-10-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

The protein encoded by this gene is a member of the SEC22 family of vesicle trafficking proteins. It is localized at the endoplasmic reticulum and it is thought to play a role in the early stages of the ER-Golgi protein trafficking. There are two alternatively spliced transcript variants encoding different isoforms described for this gene. [provided by RefSeq]

Sequence: VIFFACVVRVRDGLPLSASTDFYHTQDFLEWRRRLKSLALRLAQYPGRGSAEGCDFSIHFSSFGDV
Specifica

Specifica

NP_116752
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
33kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
VIFFACVVRVRDGLPLSASTDFYHTQDFLEWRRRLKSLALRLAQYPGRGSAEGCDFSIHFSSFGDV
RUO
SEC22C
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
9117
SEC22L3 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp761F2321/MGC13261/MGC5373/SEC22L3
SEC22C
Recombinant
wheat germ expression system
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali