missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SEMA4G Full-length ORF (AAH20960.1, 1 a.a. - 127 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00057715-P02.25ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16132943

  • 528.00€ / 25µg
Spedizione stimata: 07-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Semaphorins are a large family of conserved secreted and membrane associated proteins which possess a semaphorin (Sema) domain and a PSI domain (found in plexins, semaphorins and integrins) in the N-terminal extracellular portion. Based on sequence and structural similarities, semaphorins are put into eight classes: invertebrates contain classes 1 and 2, viruses have class V, and vertebrates contain classes 3-7. Semaphorins serve as axon guidance ligands via multimeric receptor complexes, some (if not all) containing plexin proteins. This gene encodes a class 4 semaphorin. This gene and the gene for mitochondrial ribosomal protein L43 overlap at map location 10q24.31 and are transcribed in opposite directions. [provided by RefSeq]

Sequence: MGSMSPPSAWPCVLDGPETRQDLCQPPKPCVHSHAHMEECLSAGLQCPHPHLLLVHSCFIPASGLGVPSQLPHPIWSSSPAPCGDLFVKSLGTGQPGEVRLHHSPPLPSCVALVNQPPHSPWSFSRV
Specifica

Specifica

AAH20960.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
39.9kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ20590/KIAA1619/MGC102867
SEMA4G
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
57715
SEMA4G (Human) Recombinant Protein (P02)
12.5% SDS-PAGE Stained with Coomassie Blue.
MGSMSPPSAWPCVLDGPETRQDLCQPPKPCVHSHAHMEECLSAGLQCPHPHLLLVHSCFIPASGLGVPSQLPHPIWSSSPAPCGDLFVKSLGTGQPGEVRLHHSPPLPSCVALVNQPPHSPWSFSRV
RUO
SEMA4G
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali