missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SENP5 Partial ORF (NP_689912, 2 a.a. - 109 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00205564-Q01.25ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16172437

  • 528.00€ / 25µg
Spedizione stimata: 30-10-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

The reversible posttranslational modification of proteins by the addition of small ubiquitin-like SUMO proteins (see SUMO1; MIM 601912) is required for numerous biologic processes. SUMO-specific proteases, such as SENP5, are responsible for the initial processing of SUMO precursors to generate a C-terminal diglycine motif required for the conjugation reaction. They also have isopeptidase activity for the removal of SUMO from high molecular mass SUMO conjugates (Di Bacco et al., 2006 [PubMed 16738315]).[supplied by OMIM]

Sequence: KKQRKILWRKGIHLAFSEKWNTGFGGFKKFYFHQHLCILKAKLGRPVTWNRQLRHFQGRKKALQIQKTWIKDEHLCAKTKFNVATQNVSTLSSKVKRKDAKHFISSSK
Specifica

Specifica

NP_689912
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.62kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
KKQRKILWRKGIHLAFSEKWNTGFGGFKKFYFHQHLCILKAKLGRPVTWNRQLRHFQGRKKALQIQKTWIKDEHLCAKTKFNVATQNVSTLSSKVKRKDAKHFISSSK
RUO
SENP5
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
205564
SENP5 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp564O1016/FLJ42398/MGC27076
SENP5
Recombinant
wheat germ expression system
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali