missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SERTAD3 Full-length ORF (NP_037500.2, 1 a.a. - 196 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00029946-P01.25ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16129222

  • 528.00€ / 25µg
Spedizione stimata: 07-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

The protein encoded by this gene was identified in a yeast two-hybrid assay employing the second subunit of human replication protein A as bait. It is localized to the nucleus and its expression is significantly higher in cancer cell lines compared to normal cell lines. This protein has also been shown to be a strong transcriptional co-activator. Alternative splicing has been observed at this locus and two variants, both encoding the same protein, have been identified. [provided by RefSeq]

Sequence: MVGGLKRKHSDLEEEEERWEWSPAGLQSYQQALLRISLDKVQRSLGPRAPSLRRHVLIHNTLQQLQAALRLAPAPALPPEPLFLGEEDFSLSATIGSILRELDTSMDGTEPPQNPVTPLGLQNEVPPQPDPVFLEALSSRYLGDSGLDDFFLDIDTSAVEKEPARAPPEPPHNLFCAPGSWEWNELDHIMEIILGS
Specifica

Specifica

NP_037500.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
48.2kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
RBT1
SERTAD3
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
29946
SERTAD3 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MVGGLKRKHSDLEEEEERWEWSPAGLQSYQQALLRISLDKVQRSLGPRAPSLRRHVLIHNTLQQLQAALRLAPAPALPPEPLFLGEEDFSLSATIGSILRELDTSMDGTEPPQNPVTPLGLQNEVPPQPDPVFLEALSSRYLGDSGLDDFFLDIDTSAVEKEPARAPPEPPHNLFCAPGSWEWNELDHIMEIILGS
RUO
SERTAD3
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali