missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SH3D19 Full-length ORF (AAH85613.1, 1 a.a. - 420 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00152503-P01.10ug

Additional Details : Peso : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16128733

  • 349.00€ / 10µg
Spedizione stimata: 07-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Beschreibung

Beschreibung

Sequence: MDLQKKQSNLATGLSKAKSQVFKNQDPVLPPRPKPGHPLYSKYMLSVPHGIANEDIVSQNPGELSCKRGDVLVMLKQTENNYLECQKGEDTGRVHLSQMKIITPLDEHLRSRPNDPSHAQKPVDSGAPHAVVLHDFPAEQVDDLNLTSGEIVYLLEKIDTDWYRGNCRNQIGIFPANYVKVIIDIPEGGNGKRECVSSHCVKGSRCVARFEYIGEQKDELSFSEGEIIILKEYVNEEWARGEVRGRTGIFPLNFVEPVEDYPTSGANVLSTKVPLKTKKEDSGSNSQVNSLPAEWCEALHSFTAETSDDLSFKRGDRIQILERLDSDWCRGRLQDREGIFPAVFVRPCPAEAKSMLAIVPKGRKAKALYDFRGENEDELSFKAGDIITELESVDDDWMSGELMGKSGIFPKNYIQFLQIS
Spezifikation

Spezifikation

AAH85613.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
73.5kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
EBP/EVE1/Kryn/MGC105136/MGC118910/MGC118911/MGC118912/MGC118913/SH3P19
SH3D19
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
152503
SH3D19 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MDLQKKQSNLATGLSKAKSQVFKNQDPVLPPRPKPGHPLYSKYMLSVPHGIANEDIVSQNPGELSCKRGDVLVMLKQTENNYLECQKGEDTGRVHLSQMKIITPLDEHLRSRPNDPSHAQKPVDSGAPHAVVLHDFPAEQVDDLNLTSGEIVYLLEKIDTDWYRGNCRNQIGIFPANYVKVIIDIPEGGNGKRECVSSHCVKGSRCVARFEYIGEQKDELSFSEGEIIILKEYVNEEWARGEVRGRTGIFPLNFVEPVEDYPTSGANVLSTKVPLKTKKEDSGSNSQVNSLPAEWCEALHSFTAETSDDLSFKRGDRIQILERLDSDWCRGRLQDREGIFPAVFVRPCPAEAKSMLAIVPKGRKAKALYDFRGENEDELSFKAGDIITELESVDDDWMSGELMGKSGIFPKNYIQFLQIS
RUO
SH3D19
Wheat Germ (in vitro)
GST
Liquid
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote