missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SMYD5 Full-length ORF (NP_006053.1, 1 a.a. - 302 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00010322-P01.25ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16124892

  • 528.00€ / 25µg
Spedizione stimata: 03-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Sequence: MYCSAECRLAATEQYHQVLCPGPSQDDPLHPLNKLQEAWRSIHYPPETASIMLMARMVATVKQAKDKDRWIRLFSQFCNKTANEEEEIVHKLLGDKFKGQLELLRRLFTEALYEEAVSQWFTPDGFRSLFALVGTNGQGIGTSSLSQWVHACDTLELKPQDREQLDAFIDQLYKDIEAATGEFLNCEGSGLFVLQSCCNHSCVPNAETSFPENNFLLHVTALEDIKPGEEICISYLDCCQRERSRHSRHKILRENYLFVCSCPKCLAEADEPNVTSEEEEEEEEEEEGEPEDAELGDEMTDV
Specifica

Specifica

NP_006053.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
60.8kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
NN8-4AG/RAI15/RRG1/ZMYND23
SMYD5
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
10322
SMYD5 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MYCSAECRLAATEQYHQVLCPGPSQDDPLHPLNKLQEAWRSIHYPPETASIMLMARMVATVKQAKDKDRWIRLFSQFCNKTANEEEEIVHKLLGDKFKGQLELLRRLFTEALYEEAVSQWFTPDGFRSLFALVGTNGQGIGTSSLSQWVHACDTLELKPQDREQLDAFIDQLYKDIEAATGEFLNCEGSGLFVLQSCCNHSCVPNAETSFPENNFLLHVTALEDIKPGEEICISYLDCCQRERSRHSRHKILRENYLFVCSCPKCLAEADEPNVTSEEEEEEEEEEEGEPEDAELGDEMTDV
RUO
SMYD5
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali