missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SORL1 Partial ORF (NP_003096, 82 a.a. - 181 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00006653-Q01.10ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16125675

  • 349.00€ / 10µg
Spedizione stimata: 30-10-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

This gene encodes a protein that belongs to the families of vacuolar protein sorting 10 (VPS10) domain-containing receptor proteins, of low density lipoprotein receptor (LDLR) proteins, and of fibronectin type III repeats proteins. In addition to VPS10, LDLR and fibronectin type 3 domains, this protein also includes an epidermal growth factor precursor-like module, a single transmembrane segment and a cytoplasmic tail with features similar to endocytosis- and sorting-competent receptors. Members of the VPS10 domain-containing receptor family are large with many exons but the CDS lengths are usually less than 3700 nt; this gene is an exception to the pattern with a CDS length greater than 6600 nt. Very large introns typically separate the exons encoding the VPS10 domain; the remaining exons are separated by much smaller-sized introns. The encoded protein is mainly intracellular and localizes in the paranuclear compartment. It is synthesized as a preproprotein, and when the propeptide is still attached, no binding occurs to the VPS10 domain. This gene is strongly expressed in the central nervous system. [provided by RefSeq]

Sequence: SAALQPEPIKVYGQVSLNDSHNQMVVHWAGEKSNVIVALARDSLALARPKSSDVYVSYDYGKSFKKISDKLNFGLGNRSEAVIAQFYHSPADNKRYIFAD
Specifica

Specifica

NP_003096
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
SAALQPEPIKVYGQVSLNDSHNQMVVHWAGEKSNVIVALARDSLALARPKSSDVYVSYDYGKSFKKISDKLNFGLGNRSEAVIAQFYHSPADNKRYIFAD
RUO
SORL1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
6653
SORL1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
C11orf32/FLJ21930/FLJ39258/LR11/LRP9/SORLA/SorLA-1/gp250
SORL1
Recombinant
wheat germ expression system
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali