missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SPECC1 Partial ORF (NP_690868.3, 527 a.a. - 636 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00092521-Q01.10ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Sequence: RHNNNMMAKTLEECRVTLEGLKMENGSLKSHLQGEKQKATEASAVEQTAESCEVQEMLKVARAEKDLLELSCNELRQELLKANGEIKHVSSLLAKVEKDYSYLKEICDHQSpecifica
NP_690868.3 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RHNNNMMAKTLEECRVTLEGLKMENGSLKSHLQGEKQKATEASAVEQTAESCEVQEMLKVARAEKDLLELSCNELRQELLKANGEIKHVSSLLAKVEKDYSYLKEICDHQ | |
RUO | |
CYTSB | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
92521 | |
SPECC1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ36955/HCMOGT-1/NSP/SPECC1 | |
CYTSB | |
Recombinant | |
wheat germ expression system |