missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human TEX13B Full-length ORF (NP_112563.1, 1 a.a. - 312 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00056156-P01.10ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
This gene is similar to a mouse gene that is expressed in the testis. [provided by RefSeq]
Sequence: MALRPEDPSSGFRHGNVVAFIIEKMARHTKGPEFYFENISLSWEEVEDKLRAILEDSEVPSEVKEACTWGSLALGVRFAHRQGQLQNRRVQWLQGFAKLHRSAALVLASNLTELKEQQEMECNEATFQLQLTETSLAEVQRERDMLRWKLFHAELAPPQGQGQATVFPGLATAGGDWTEGAGEQEKEAVAAAGAAGGKGEERYAEAGPAPAEVLQGLGGGFRQPLGAIVAGKLHLCGAEGERSQVSTNSHVCLLWAWVHSLTGASSCPAPYLIHILIPMPFVRLLSHTQYTPFTSKGHRTGSNSDAFQLGGLSpecifica
NP_112563.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
60.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
TGC3B/TSGA5 | |
TEX13B | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
56156 | |
TEX13B (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MALRPEDPSSGFRHGNVVAFIIEKMARHTKGPEFYFENISLSWEEVEDKLRAILEDSEVPSEVKEACTWGSLALGVRFAHRQGQLQNRRVQWLQGFAKLHRSAALVLASNLTELKEQQEMECNEATFQLQLTETSLAEVQRERDMLRWKLFHAELAPPQGQGQATVFPGLATAGGDWTEGAGEQEKEAVAAAGAAGGKGEERYAEAGPAPAEVLQGLGGGFRQPLGAIVAGKLHLCGAEGERSQVSTNSHVCLLWAWVHSLTGASSCPAPYLIHILIPMPFVRLLSHTQYTPFTSKGHRTGSNSDAFQLGGL | |
RUO | |
TEX13B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |