missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TM2D3 Full-length ORF (AAH08873.1, 1 a.a. - 221 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00080213-P01.10ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16124843

  • 349.00€ / 10µg
Spedizione stimata: 26-10-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

The protein encoded by this gene contains a structural module related to that of the seven transmembrane domain G protein-coupled receptor superfamily. This protein has sequence and structural similarities to the beta-amyloid binding protein (BBP), but, unlike BBP, it does not regulate a response to beta-amyloid peptide. This protein may have regulatory roles in cell death or proliferation signal cascades. Several alternatively spliced transcript variants of this gene are described but the full length nature of some variants has not been determined. Multiple polyadenylation sites have been found in this gene. [provided by RefSeq]

Sequence: MAGGVRPLRGLRALCRVLLFLSQFCILSGGESTEIPPYVMKCPSNGLCSRLPADCIDCTTNFSCTYGKPVTFDCAVKPSVTCVDQDFKSQKNFIINMTCRFCWQLPETDYECTNSTSCMTVSCPRQRYPANCTVRDHVHCLGNRTFPKMLYCNWTGGYKWSTALALSITLGGFGADRFYLGQWREGLGKLFSFGGLGIWTLIDVLLIGVGYVGPADGSLYI
Specifica

Specifica

AAH08873.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
50.8kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BLP2
TM2D3
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
80213
TM2D3 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAGGVRPLRGLRALCRVLLFLSQFCILSGGESTEIPPYVMKCPSNGLCSRLPADCIDCTTNFSCTYGKPVTFDCAVKPSVTCVDQDFKSQKNFIINMTCRFCWQLPETDYECTNSTSCMTVSCPRQRYPANCTVRDHVHCLGNRTFPKMLYCNWTGGYKWSTALALSITLGGFGADRFYLGQWREGLGKLFSFGGLGIWTLIDVLLIGVGYVGPADGSLYI
RUO
TM2D3
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali