missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human TNKS Partial ORF (NP_003738, 1017 a.a. - 1124 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00008658-Q01.25ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Sequence: TERKEGEVAGLDMNISQFLKSLGLEHLRDIFETEQITLDVLADMGHEELKEIGINAYGHRHKLIKGVERLLGGQQGTNPYLTFHCVNQGTILLDLAPEDKEYQSVEEESpecifica
NP_003738 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.62kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
TERKEGEVAGLDMNISQFLKSLGLEHLRDIFETEQITLDVLADMGHEELKEIGINAYGHRHKLIKGVERLLGGQQGTNPYLTFHCVNQGTILLDLAPEDKEYQSVEEE | |
RUO | |
TNKS | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
8658 | |
TNKS (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PARP-5a/PARP5A/PARPL/TIN1/TINF1/TNKS1 | |
TNKS | |
Recombinant | |
wheat germ expression system |