missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TPRX1 Partial ORF (NP_940881.2, 313 a.a. - 411 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00284355-Q01.10ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16172737

  • 349.00€ / 10µg
Spedizione stimata: 26-10-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Homeobox genes encode DNA-binding proteins, many of which are thought to be involved in early embryonic development. Homeobox genes encode a DNA-binding domain of 60 to 63 amino acids referred to as the homeodomain. This gene is a member of the TPRX homeobox gene family. [provided by RefSeq]

Sequence: GSGSVPAPIPGPGSLPAPAPLWPQSPDASDFLPDTQLFPHFTELLLPLDPLEGSSVSTMTSQYQEGDDSMGKKHSGSQPQEEGGSVNENHSGPRLLLDL
Specifica

Specifica

NP_940881.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue
GSGSVPAPIPGPGSLPAPAPLWPQSPDASDFLPDTQLFPHFTELLLPLDPLEGSSVSTMTSQYQEGDDSMGKKHSGSQPQEEGGSVNENHSGPRLLLDL
RUO
TPRX1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
284355
TPRX1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ40321/TPRX
TPRX1
Recombinant
wheat germ expression system
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali