missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TRIM31 Partial ORF (NP_008959, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00011074-Q01.10ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16126132

  • 349.00€ / 10µg
Spedizione stimata: 07-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to both the cytoplasm and the nucleus. Its function has not been identified. [provided by RefSeq]

Sequence: MASGQFVNKLQEEVICPICLDILQKPVTIDCGHNFCLKCITQIGETSCGFFKCPLCKTSVRKNAIRFNSLLRNLVEKIQALQASEVQSKRKEATCPRHQE
Specifica

Specifica

NP_008959
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MASGQFVNKLQEEVICPICLDILQKPVTIDCGHNFCLKCITQIGETSCGFFKCPLCKTSVRKNAIRFNSLLRNLVEKIQALQASEVQSKRKEATCPRHQE
RUO
TRIM31
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
11074
TRIM31 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
C6orf13/HCG1/HCGI/RNF
TRIM31
Recombinant
wheat germ expression system
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali