missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TRPA1 Partial ORF (NP_015628, 1033 a.a. - 1117 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00008989-Q01.25ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16129705

  • 528.00€ / 25µg
Spedizione stimata: 02-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

The structure of the protein encoded by this gene is highly related to both the protein ankyrin and transmembrane proteins. The specific function of this protein has not yet been determined; however, studies indicate the function may involve a role in signal transduction and growth control. [provided by RefSeq]

Sequence: IPNADKSLEMEILKQKYRLKDLTFLLEKQHELIKLIIQKMEIISETEDDDSHCSFQDRFKKEQMEQRNSRWNTVLRAVKAKTHHL
Specifica

Specifica

NP_015628
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.09kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
IPNADKSLEMEILKQKYRLKDLTFLLEKQHELIKLIIQKMEIISETEDDDSHCSFQDRFKKEQMEQRNSRWNTVLRAVKAKTHHL
RUO
TRPA1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
8989
TRPA1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ANKTM1
TRPA1
Recombinant
wheat germ expression system
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali