missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TTC23 Full-length ORF (AAH15728.1, 1 a.a. - 393 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00064927-P01.25ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16173813

  • 508.00€ / 25µg
Spedizione stimata: 26-08-2024
per visualizzare lo stock disponibile

Descrizione

Descrizione

Sequence: MQESQETHISNHLDEVVAAVSITHRKKFQNKLLQTALFQPPREKLHLCEEKAKSYSNSHEYKQAVHELVRCVALTRICYGDSHWKLAEAHVNLAQGYLQLKGLSLQAKQHAEKARQILANSIVPPYSENTDVFKFSIELFHTMGRALLSLQKFKEAAENLTKAERLSKELLQCGRIIKEEWIEIEARIRLSFAQVYQGQKKSKEALSHYQAALEYVEISKGETSRECVPILRELAGVEQALGLHDVSINHFLQAHLIILSRSPSQVEAADSAHIVAHAAVASGRHEHHDVAEQYFQESMAHLKDSEGMGRTKFLSIQDEFCHFLQMTGQKERATSILRESLEAKVEAFGDFSPEVAETYRLLGGADLAQGNHSGARKKLKKGDLSFLLSATLP
Specifica

Specifica

AAH15728.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
70.6kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ12572/FLJ12939/HCC-8
TTC23
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
64927
TTC23 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MQESQETHISNHLDEVVAAVSITHRKKFQNKLLQTALFQPPREKLHLCEEKAKSYSNSHEYKQAVHELVRCVALTRICYGDSHWKLAEAHVNLAQGYLQLKGLSLQAKQHAEKARQILANSIVPPYSENTDVFKFSIELFHTMGRALLSLQKFKEAAENLTKAERLSKELLQCGRIIKEEWIEIEARIRLSFAQVYQGQKKSKEALSHYQAALEYVEISKGETSRECVPILRELAGVEQALGLHDVSINHFLQAHLIILSRSPSQVEAADSAHIVAHAAVASGRHEHHDVAEQYFQESMAHLKDSEGMGRTKFLSIQDEFCHFLQMTGQKERATSILRESLEAKVEAFGDFSPEVAETYRLLGGADLAQGNHSGARKKLKKGDLSFLLSATLP
RUO
TTC23
Wheat Germ (in vitro)
GST
Liquid
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali