missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human VPS41 Full-length ORF (P49754, 1 a.a. - 854 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00027072-P01.25ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16128522

  • 528.00€ / 25µg
Spedizione stimata: 26-10-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Vesicle mediated protein sorting plays an important role in segregation of intracellular molecules into distinct organelles. Genetic studies in yeast have identified more than 40 vacuolar protein sorting (VPS) genes involved in vesicle transport to vacuoles. This gene encodes the human ortholog of yeast Vps41 protein which is also conserved in Drosophila, tomato, and Arabidopsis. Expression studies in yeast and human indicate that this protein may be involved in the formation and fusion of transport vesicles from the Golgi. Several transcript variants encoding different isoforms have been described for this gene, however, the full-length nature of not all is known. [provided by RefSeq]

Sequence: MAEAVEQETGSLEESTDESEEEESEEEPKLKYERLSNGVTEILQKDAASCMTVHDKFLALGTHYGKVYLLDVQGNITQKFDVSPVKINQISLDESGEHMGVCSEDGKVQVFGLYSGEEFHETFDCPIKIIAVHPHFVRSSCKQFVTGGKKLLLFERSWMNRWKSAVLHEGEGNIRSVKWRGHLIAWANNMGVKIFDIISKQRITNVPRDDISLRPDMYPCSLCWKDNVTLIIGWGTSVKVCSVKERHASEMRDLPSRYVEIVSQFETEFYISGLAPLCDQLVVLSYVKEISEKTEREYCARPRLDIIQPLSETCEEISSDALTVRGFQENECRDYHLEYSEGESLFYIVSPRDVVVAKERDQDDHIDWLLEKKKYEEALMAAEISQKNIKRHKILDIGLAYINHLVERGDYDIAARKCQKILGKNAALWEYEVYKFKEIGQLKAISPYLPRGDPVLKPLIYEMILHEFLESDYEGFATLIREWPGDLYNNSVIVQAVRDHLKKDSQNKTLLKTLAELYTYDKNYGNALEIYLTLRHKDVFQLIHKHNLFSSIKDKIVLLMDFDSEKAVDMLLDNEDKISIKKVVEELEDRPELQHVYLHKLFKRDHHKGQRYHEKQISLYAEYDRPNLLPFLRDSTHCPLEKALEICQQRNFVEETVYLLSRMGNSRSALKMIMEELHDVDKAIEFAKEQDDGELWEDLILYSIDKPPFITGLLNNIGTHVDPILLIHRIKEGMEIPNLRDSLVKILQDYNLQILLREGCKKILVADSLSLLKKMHRTQMKGVLVDEENICESCLSPILPSDAAKPFSVVVFHCRHMFHKECLPMPSMNSAAQFCNICSAKNRGPGSAILEMKK
Specifica

Specifica

P49754
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
124.9kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HVPS41/HVSP41/hVps41p
VPS41
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
27072
VPS41 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAEAVEQETGSLEESTDESEEEESEEEPKLKYERLSNGVTEILQKDAASCMTVHDKFLALGTHYGKVYLLDVQGNITQKFDVSPVKINQISLDESGEHMGVCSEDGKVQVFGLYSGEEFHETFDCPIKIIAVHPHFVRSSCKQFVTGGKKLLLFERSWMNRWKSAVLHEGEGNIRSVKWRGHLIAWANNMGVKIFDIISKQRITNVPRDDISLRPDMYPCSLCWKDNVTLIIGWGTSVKVCSVKERHASEMRDLPSRYVEIVSQFETEFYISGLAPLCDQLVVLSYVKEISEKTEREYCARPRLDIIQPLSETCEEISSDALTVRGFQENECRDYHLEYSEGESLFYIVSPRDVVVAKERDQDDHIDWLLEKKKYEEALMAAEISQKNIKRHKILDIGLAYINHLVERGDYDIAARKCQKILGKNAALWEYEVYKFKEIGQLKAISPYLPRGDPVLKPLIYEMILHEFLESDYEGFATLIREWPGDLYNNSVIVQAVRDHLKKDSQNKTLLKTLAELYTYDKNYGNALEIYLTLRHKDVFQLIHKHNLFSSIKDKIVLLMDFDSEKAVDMLLDNEDKISIKKVVEELEDRPELQHVYLHKLFKRDHHKGQRYHEKQISLYAEYDRPNLLPFLRDSTHCPLEKALEICQQRNFVEETVYLLSRMGNSRSALKMIMEELHDVDKAIEFAKEQDDGELWEDLILYSIDKPPFITGLLNNIGTHVDPILLIHRIKEGMEIPNLRDSLVKILQDYNLQILLREGCKKILVADSLSLLKKMHRTQMKGVLVDEENICESCLSPILPSDAAKPFSVVVFHCRHMFHKECLPMPSMNSAAQFCNICSAKNRGPGSAILEMKK
RUO
VPS41
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali