missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human WFDC5 Partial ORF (NP_663627, 25 a.a. - 123 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00149708-Q01.25ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16172107

  • 508.00€ / 25µg
Spedizione stimata: 26-08-2024
per visualizzare lo stock disponibile

Descrizione

Descrizione

This gene encodes a member of the WAP-type four-disulfide core (WFDC) domain family. Most WFDC proteins contain only one WFDC domain, and this encoded protein contains two WFDC domains. The WFDC domain, or WAP signature motif, contains eight cysteines forming four disulfide bonds at the core of the protein, and functions as a protease inhibitor. Most WFDC gene members are localized to chromosome 20q12-q13 in two clusters: centromeric and telomeric. This gene belongs to the centromeric cluster. [provided by RefSeq]

Sequence: RKKGEKSGGCPPDDGPCLLSVPDQCVEDSQCPLTRKCCYRACFRQCVPRVSVKLGSCPEDQLRCLSPMNHLCHKDSDCSGKKRCCHSACGRDCRDPARG
Specifica

Specifica

NP_663627
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
RKKGEKSGGCPPDDGPCLLSVPDQCVEDSQCPLTRKCCYRACFRQCVPRVSVKLGSCPEDQLRCLSPMNHLCHKDSDCSGKKRCCHSACGRDCRDPARG
RUO
WFDC5
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
149708
WFDC5 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
PRG5/WAP1/dJ211D12.5
WFDC5
Recombinant
wheat germ expression system
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali