missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ZKSCAN1 Full-length ORF (AAH22378.1, 1 a.a. - 563 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00007586-P02.25ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Sequence: MMTAESREATGLSPQAAQEKDGIVIAKVEEEDEEDHMWGQDSTLQDTPPPDPEIFRQRFRRFCYQNTFGPREALSRLKELCHQWLRPEINTKEQILELLVLEQFLSILPKELQVWLQEYRPDSGEEAVTLLEDLELDLSGQQVPGQVHGPEMLARGMVPLDPVQESSSFDLHHEATQSHFKHSSRKPRLLQSRALPAAHIPAPPHEGSPRDQAMASALFTADSQAMVKIEDMAVSLILEEWGCQNLARRNLSRDNRQENYGSAFPQGGENRNENEESTSKAETSEDSASRGETTGRSQKEFGEKRDQEGKTGERQQKNPEEKTRKEKRDSGPAIGKDKKTITGERGPREKGKGLGRSFSLSSNFTTPEEVPTGTKSHRCDECGKCFTRSSSLIRHKIIHTGEKPYECSECGKAFSLNSNLVLHQRIHTGEKPHECNECGKAFSHSSNLILHQRIHSGEKPYECNECGKAFSQSSDLTKHQRIHTGEKPYECSECGKAFNRNSYLILHRRIHTREKPYKCTKCGKAFTRSSTLTLHHRIHARERASEYSPASLDAFGAFLKSCVSpecifica
AAH22378.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
90kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
9130423L19Rik/KOX18/MGC138429/PHZ-37/ZNF139/ZNF36 | |
ZKSCAN1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
7586 | |
ZKSCAN1 (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MMTAESREATGLSPQAAQEKDGIVIAKVEEEDEEDHMWGQDSTLQDTPPPDPEIFRQRFRRFCYQNTFGPREALSRLKELCHQWLRPEINTKEQILELLVLEQFLSILPKELQVWLQEYRPDSGEEAVTLLEDLELDLSGQQVPGQVHGPEMLARGMVPLDPVQESSSFDLHHEATQSHFKHSSRKPRLLQSRALPAAHIPAPPHEGSPRDQAMASALFTADSQAMVKIEDMAVSLILEEWGCQNLARRNLSRDNRQENYGSAFPQGGENRNENEESTSKAETSEDSASRGETTGRSQKEFGEKRDQEGKTGERQQKNPEEKTRKEKRDSGPAIGKDKKTITGERGPREKGKGLGRSFSLSSNFTTPEEVPTGTKSHRCDECGKCFTRSSSLIRHKIIHTGEKPYECSECGKAFSLNSNLVLHQRIHTGEKPHECNECGKAFSHSSNLILHQRIHSGEKPYECNECGKAFSQSSDLTKHQRIHTGEKPYECSECGKAFNRNSYLILHRRIHTREKPYKCTKCGKAFTRSSTLTLHHRIHARERASEYSPASLDAFGAFLKSCV | |
RUO | |
ZKSCAN1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |