missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ZNF585B Full-length ORF (AAH93625.1, 1 a.a. - 769 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00092285-P01.10ug

 vedi altre versioni di questo prodotto

Codice prodotto. 16126723

  • 335.00€ / 10µg

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Esplora altre offerte speciali
Questo articolo non è restituibile. Consulta la politica di reso

Descrizione

Descrizione

Sequence: MPASWTSPQKSSALAPEDHGSSYEGSVSFRDVAIDFSREEWRHLDLSQRNLYRDVMLETYSHLLSVGYQVPKPEVVMLEQGKEPWALQGERPRHSCPGEKLWDHNQHRKIIGYKPASSQDQKIYSGEKSYECAEFGKSFTWKSQFKVHLKVPTGEKLYVCIECGRAFVQKPEFITHQKTHMREKPYKCNECGKSFFQVSSLFRHHRIHTGEKLYECSECGKGFPYNSDLSIHEKIHTGERHHECTDCGKAFTQKSTLKIHQKIHTGERSYICIECGQAFIQKTQLIAHRRIHSGEKPYECNNCGKSFISKSQLQVHQRVHTRVKPYICTEYGKVFSNNSNLITHEKIQSREKSSICTECGKAFTYRSELIIHQRIHTGEKPYECSDCGRAFTQKSALTVHQRIHTGEKSYICMKCGLAFIRKAHLITHQIIHTGEKPYKCGHCGKLFTSKSQLHVHKRIHTGEKPYVCNKCGKAFTNRSNLITHQKTHTGEKSYICSKCGKAFTQRSDLITHQRIHTGEKPYECNTCGKAFTQKSNLNIHQKIHTGERQYECHECGKAFNQKSILIVHQKIHTGEKPYVCTECGRAFIRKSNFITHQRIHTGEKPYECSDCGKSFTSKSQLLVHQPVHTGEKPYVCAECGKAFSGRSNLSKHQKTHTGEKPYICSECGKTFRQKSELITHHRIHTGEKPYECSDCGKSFTKKSQLQVHQRIHTGEKPYVCAECGKAFSNRSNLNKHQTTHTGDKPYKCGICGKGFVQKSVFSVHQSSHA
Specifica

Specifica

AAH93625.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
114.5kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ14928/SZFP41
ZNF585B
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
92285
ZNF585B (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MPASWTSPQKSSALAPEDHGSSYEGSVSFRDVAIDFSREEWRHLDLSQRNLYRDVMLETYSHLLSVGYQVPKPEVVMLEQGKEPWALQGERPRHSCPGEKLWDHNQHRKIIGYKPASSQDQKIYSGEKSYECAEFGKSFTWKSQFKVHLKVPTGEKLYVCIECGRAFVQKPEFITHQKTHMREKPYKCNECGKSFFQVSSLFRHHRIHTGEKLYECSECGKGFPYNSDLSIHEKIHTGERHHECTDCGKAFTQKSTLKIHQKIHTGERSYICIECGQAFIQKTQLIAHRRIHSGEKPYECNNCGKSFISKSQLQVHQRVHTRVKPYICTEYGKVFSNNSNLITHEKIQSREKSSICTECGKAFTYRSELIIHQRIHTGEKPYECSDCGRAFTQKSALTVHQRIHTGEKSYICMKCGLAFIRKAHLITHQIIHTGEKPYKCGHCGKLFTSKSQLHVHKRIHTGEKPYVCNKCGKAFTNRSNLITHQKTHTGEKSYICSKCGKAFTQRSDLITHQRIHTGEKPYECNTCGKAFTQKSNLNIHQKIHTGERQYECHECGKAFNQKSILIVHQKIHTGEKPYVCTECGRAFIRKSNFITHQRIHTGEKPYECSDCGKSFTSKSQLLVHQPVHTGEKPYVCAECGKAFSGRSNLSKHQKTHTGEKPYICSECGKTFRQKSELITHHRIHTGEKPYECSDCGKSFTKKSQLQVHQRIHTGEKPYVCAECGKAFSNRSNLNKHQTTHTGDKPYKCGICGKGFVQKSVFSVHQSSHA
RUO
ZNF585B
Wheat Germ (in vitro)
GST
Liquid
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali