missing translation for 'onlineSavingsMsg'
Learn More

IGBP1 Mouse anti-Human, Mouse, Clone: 2D11A8, Proteintech

Mouse Monoclonal Antibody

Marca:  Proteintech 60256-1-IG-150UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16897295

  • 349.00€ / 150µL

Spedizione stimata: 16-12-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

The proliferation and differentiation of B cells is dependent upon a B-cell antigen receptor (BCR) complex. Binding of antigens to specific B-cell receptors results in a tyrosine phosphorylation reaction through the BCR complex and leads to multiple signal transduction pathways.
Specifica

Specifica

IGBP1
Monoclonal
1.1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P78318, Q61249
IGBP1
IGBP1 Fusion Protein Ag6782
150 μL
Primary
Mouse, Human
Antibody
IgG1
Western Blot, Immunohistochemistry (Paraffin)
2D11A8
Unconjugated
IGBP1
ALPHA 4, CD79a binding protein 1, IBP1, IGBP1, Protein alpha 4
Mouse
Protein G
RUO
18518, 3476
-20°C
Liquid
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali