missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-10 Rat anti-Human, Virus, APC-R700, Clone: JES3-19F1, BD Biosciences™
Rat Monoclonal Antibody
Marca: BD Biosciences 567013
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
IL-10 Monoclonal antibody specifically detects IL-10 in Human, Virus samples. It is validated for Flow Cytometry
Specifica
| IL-10 | |
| Monoclonal | |
| APC-R700 | |
| Interleukin-10; CSIF; Cytokine synthesis inhibitory factor; TGIF | |
| Recombinant Human IL-10 | |
| RUO | |
| Human, Virus | |
| IgG2a κ |
| Flow Cytometry | |
| JES3-19F1 | |
| Aqueous buffered solution containing BSA, protein stabilizer, glycerol and ≤0.09% sodium azide. | |
| Rat | |
| 25 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto