missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-22 Mouse anti-Human, Alexa Fluor 647, Clone: MH22B2, BD Biosciences™
Mouse Monoclonal Antibody
Marca: BD Biosciences 567160
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
IL-22 Monoclonal antibody specifically detects IL-22 in Human samples. It is validated for Intracellular staining (flow cytometry)
Specifica
| IL-22 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 0.1 mg | |
| Primary | |
| The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to Alexa Fluor 647 under optimum conditions, and unreacted Alexa Fluor 647 was removed. Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| MH22B2 | |
| Alexa Fluor 647 | |
| interleukin-22; IL-10-related T-cell-derived inducible factor; IL-TIF; ILTIF; IL-22a, ilTIFa; Iltif; TIFa | |
| Human IL-22 Recombinant Protein | |
| RUO | |
| Human | |
| IgG2a κ |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto