missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-33R (ST2) Recombinant Rabbit Monoclonal Antibody (ST2IL33R-H2), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marca: Thermo Scientific MA537204
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
IL-33R (ST2) Recombinant Monoclonal antibody specifically detects IL-33R (ST2) in Human samples. It is validated for Flow Cytometry.Specifica
IL-33R (ST2) | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.20% BSA and 0.09% sodium azide | |
Q01638 | |
IL1RL1 | |
human recombinant ST2-Fc. | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG κ |
Flow Cytometry | |
ST2IL33R-H2 | |
Unconjugated | |
IL1RL1 | |
DER4; Fit1; Fit-1; Fos-responsive gene 1 protein; growth stimulation-expressed; homolog of mouse growth stimulation-expressed; IL-1 R4; IL1RL1; IL-1RL1; IL33R; IL-33R; interleukin 1 receptor like 1; interleukin 1 receptor-like 1; interleukin 1 receptor-related protein; Interleukin1 receptor like 1; interleukin-1 receptor-like 1; Interleukin-33 receptor alpha chain; Ly84; lymphocyte antigen 84; Protein ST2; Protein T1; sIL 33R; soluble IL 33 Receptor; soluble IL 33R; St2; ST2L; St2-rs1; ST2V; Ste2; T1; T1/ST2 | |
Rabbit | |
Protein A/G | |
RUO | |
9173 | |
-20°C | |
Liquid |