missing translation for 'onlineSavingsMsg'
Learn More

IL-4 Mouse anti-Human, Clone: 1G2C5, Proteintech

Mouse Monoclonal Antibody

Marca:  Proteintech 66142-1-IG-20UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16897745

  • 121.38€ / 20µL

Spedizione stimata: 11-10-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

Interleukin-4 (IL-4) is a pleiotropic, immune-modulatory cytokine produced by activated T cells, and serves as a ligand for interleukin 4 receptor. IL-4 has been found to have the broadest range of actions. The interleukin 4 receptor also binds to IL13, which may contribute to many overlapping functions of this cytokine and IL13. STAT6, a signal transducer and activator of transcription, has been shown to play an important role in mediating the immune regulatory signal of IL-4. The IL-4 gene, IL-3, IL-5, IL-13, and CSF2 form a cytokine gene cluster on chromosome 5q. In particular, IL-4 is located in close proximity to IL13 on the chromosome. Further, the IL-4 gene, IL13 and IL5 are found to be coordinated by several long-range regulatory elements over a 120 kilobase range on the chromosome. IL-4 delta 2 is a known splice variant of IL-4.
Specifica

Specifica

IL-4
Monoclonal
0.74 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P05112
IL4
IL-4 Fusion Protein Ag9940
20 μL
Primary
Human
Antibody
IgG2a
Immunohistochemistry (Paraffin), Western Blot
1G2C5
Unconjugated
IL4
B cell stimulatory factor 1, Binetrakin, BSF 1, BSF1, IL 4, IL4, IL-4, interleukin 4, PS1TP5BP1
Mouse
Protein G
RUO
3565
-20°C
Liquid
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali