missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL13RA2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11059-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
The protein encoded by this gene is closely related to Il13RA1, a subuint of the interleukin 13 receptor complex. This protein binds IL13 with high affinity, but lacks cytoplasmic domain, and does not appear to function as a signal mediator. It is reported to play a role in the internalization of IL13.Specifica
IL13RA2 | |
Polyclonal | |
Unconjugated | |
IL13RA2 | |
CD213A2, CT19, IL 13 receptor subunit alpha 2, IL 13R, IL 13R alpha 2, IL 13R subunit alpha 2, IL 13RA2, IL13BP, IL13R, IL13RA2, IL13Ralpha2, Interleukin 13 binding protein | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
16165, 171060, 3598 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O88786, Q14627, Q8VHK6 | |
IL13RA2 | |
IL13RA2 Fusion Protein Ag1534 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |