missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL17RE Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11984-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
The interleukin 17 receptor E (IL-17RE) shares limited similarity to the receptor of IL-17A, a ubiquitous type I membrane glycoprotein that with IL-17A plays a pathogenic role in many inflammatory and autoimmune diseases such as rheumatoid arthritis. IL-17RE is the functional receptor of IL-17C and has been suggested to mediate mucosal immunity to infection with intestinal pathogens. IL-17C is produced by epithelia in response to bacterial challenge and binds to a receptor complex formed by IL-17RE and IL-17RA on the epithelial cell surface, thereby regulating the immune function of epithelial cells in an autocrine manner.Specifica
IL17RE | |
Polyclonal | |
Unconjugated | |
IL17RE | |
IL 17 receptor E, IL 17RE, IL17RE, interleukin 17 receptor E | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
132014, 57890 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8BH06, Q8NFR9 | |
Il17re | |
IL17RE Fusion Protein Ag2612 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |