Learn More
IL2RG Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11409-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
IL2RG is an important signaling component of many interleukin receptors, including those of interleukin -2, -4, -7 and -21, and is thus referred to as the common gamma chain. Mutations in this gene cause X-linked severe combined immunodeficiency (XSCID), as well as X-linked combined immunodeficiency (XCID), a less severe immunodeficiency disorder.Specifica
IL2RG | |
Polyclonal | |
Unconjugated | |
IL2RG | |
CD132, IL 2 receptor subunit gamma, IL 2R subunit gamma, IL 2RG, IL2RG, IMD4, p64, SCIDX, SCIDX1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
140924, 16186, 3561 | |
-20°C | |
Liquid |
Flow Cytometry, Western Blot | |
0.14 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P31785, P34902 | |
IL2RG | |
IL2RG Fusion Protein Ag1911 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |