Learn More
Motic™ Images Assembly Professional Edition (for PC only)
Get the most from your Motic™ microscope system with purposeful and thoughtfully designed software from Motic™. Motic™ Images Assembly Professional Edition (for PC only) is a one-stop software tool for combining up to 400 images to create single, high-magnification images with a large field of view.
Marca: Motic™ SE990639
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
- Motic™ Images Assembly Pro will combine up to 400 images (20X20) images taken in one plane.
- The complex software algorithm will take into account overlap and shifting to produce high-magnification images with a large field of view.
- Motic™ Images Assembly Pro is contained within the Motic™ Images Advanced package and is also available as a separate purchase.
- Windows™ XP or 2000 are recommended to run this software.
- Compatible with Motic™ Images Plus 2.0 and integrated into Motic™ Advanced 3.2
Specifica
Software | |
Image stitching software | |
Windows™ XP or 2000 | |
Microscope/camera/computer |