Learn More
Novus Biologicals™ Imiquimod, TLR7 Ligand
Applications: Functional, In vitro assay, Ligand Activation
Marca: Novus Biologicals™ NBP2-26228-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
Imiquimod, TLR7 ligand is an imidazoquinoline amine analog to guanosine. As a synthetic molecule of the Imidazoquinoline family, it has potent immunostimulatory activity. Imiquimod has been shown to activate only TLR7, this activation is MyD88 dependant and leads to the induction of the transcription factor NF-kB.
- Species: Human, Mouse
- Specificity: Imiquimod is human/mouse TLR7 agonist
- Stimulation of TLR7 can be achieved with 5-10μg/mL
Recommended Storage
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Specifica
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
For activation of TLR7 | |
Imiquimod, TLR7 Ligand | |
1 mg |