Learn More
Masterflex™ In-Line 304 Stainless Steel Low-Profile Strainer Mesh
Ideal for small-scale low-volume filtration. Use low-profile strainers in research applications, cleaning equipment and laboratory equipment.
Marca:  Masterflex™ 29595-33
Dettagli aggiuntivi : Peso : 0.80000kg
Codice prodotto. 11730569
  • 14.80€ / Pezzo
Spedizione stimata: 04-07-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
  • Build your own economical filtration system to filter out particulates from your fluids. Available in three classes: low-profile, intermediate and large. Modify or change components of the same class to meet the demands of your application.
  • REQUIRED SYSTEM COMPONENTS
  • Order four components from the same class:
  • Strainer top
  • Strainer mesh
  • Strainer bottom
  • Gasket
Specifica
Specifica
Stainless Steel
480 Micron Strainer Mesh, 40 x 36