Learn More
Masterflex™ In-Line 304 Stainless Steel Low-Profile Strainer Mesh
Ideal for small-scale low-volume filtration. Use low-profile strainers in research applications, cleaning equipment and laboratory equipment.
Marca: Masterflex™ 29595-33
Dettagli aggiuntivi : Peso : 0.80000kg
Descrizione
- Build your own economical filtration system to filter out particulates from your fluids. Available in three classes: low-profile, intermediate and large. Modify or change components of the same class to meet the demands of your application.
- REQUIRED SYSTEM COMPONENTS
- Order four components from the same class:
- Strainer top
- Strainer mesh
- Strainer bottom
- Gasket
Specifica
Stainless Steel | |
480 Micron Strainer Mesh, 40 x 36 |