Learn More
Smeg™ Injection Trolley for Laboratory Glassware Washers
Use these trolleys with Smeg™ glassware washers. Smeg Injection Trolleys for Laboratory Glassware Washers are available in a variety of configurations to fit a wide range of applications.
Marca: Smeg™ LM40S
Dettagli aggiuntivi : Peso : 0.80000kg
Descrizione
- For use with Smeg glassware washers
- Available with one, two and mixed spraying/injection levels
- Clean and sanitize a wide variety of labware
Specifica
Stainless Steel | |
40 items | |
Injection trolley with two spraying/injection levels | |
Laboratory glassware washers | |
1 Each |
Injection Trolley | |
Lower level with rotary sprayers to hold various supports (beakers, test tubes, flasks) and an upper level with a 40-position injection trolley | |
Simultaneous washing of narrow-necked and other types of glassware | |
40 U4140 spigots, without drying system connection |