Learn More
Smeg™ Injection Trolley for Laboratory Glassware Washers
Use these trolleys with Smeg™ glassware washers. Smeg Injection Trolleys for Laboratory Glassware Washers are available in a variety of configurations to fit a wide range of applications.
Marca:  Smeg™ LM80
Dettagli aggiuntivi : Peso : 0.80000kg
 vedi altre versioni di questo prodotto
Codice prodotto. 11945564
  • 3315.00€ / Pezzo
Spedizione stimata: 24-06-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
  • For use with Smeg glassware washers
  • Available with one, two and mixed spraying/injection levels
  • Clean and sanitize a wide variety of labware
Specifica
Specifica
Stainless Steel
68 items
Injection trolley with two injection levels
Laboratory glassware washers
1 Each
Injection Trolley
two injection levels for a capacity of 68 items.
Washing up to 68 glassware items
Without drying system connection