Learn More
Smeg™ Injection Trolley for Laboratory Glassware Washers
Use these trolleys with Smeg™ glassware washers. Smeg Injection Trolleys for Laboratory Glassware Washers are available in a variety of configurations to fit a wide range of applications.
Marca: Smeg™ LPM20/20
Dettagli aggiuntivi : Peso : 1.00000kg
Descrizione
- For use with Smeg glassware washers
- Available with one, two and mixed spraying/injection levels
- Clean and sanitize a wide variety of labware
Specifica
Stainless Steel | |
Laboratory glassware washers | |
1 Each |
20 narrow-necked items, plus 20 pipettes | |
Without drying system connection |