missing translation for 'onlineSavingsMsg'
Learn More
Learn More
iNOS Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 4 publications
Marca: Novus Biologicals NBP1-97471-0.05mg
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
iNOS Polyclonal specifically detects iNOS in Mouse, Rat, Bovine samples. It is validated for Western Blot, Immunoprecipitation.
Specifica
| iNOS | |
| Polyclonal | |
| Unconjugated | |
| PBS and 50% Glycerol with 0.09% Sodium Azide | |
| NOS2 | |
| Synthetic peptide corresponding to aa 1131-1144 of mouse iNOS. | |
| Protein A purified | |
| RUO | |
| 4843 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| 1.0 mg/mL | |
| Western Blot 1:1000, Immunoprecipitation 1:10-1:500 | |
| EC 1.14.13.39, Hepatocyte NOS, HEP-NOSPeptidyl-cysteine S-nitrosylase NOS2, Inducible NO synthase, Inducible NOS, INOS, nitric oxide synthase 2, inducible, nitric oxide synthase 2A (inducible, hepatocytes), nitric oxide synthase, inducible, nitric oxide synthase, macrophage, NOS, NOS, type II, NOS2ANOS type II | |
| Rabbit | |
| 131 kDa | |
| 0.05 mg | |
| Primary | |
| Mouse, Rat, Bovine | |
| Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto