Learn More
Hirschmann™ Intake Valves
For use with ceramus™, solarus™, opus™, ceramus™ classic for 30 + 60 ml, EM dispenser for 10-60 ml akku drive™
Marca: Hirschmann™ 9316510
Dettagli aggiuntivi : Peso : 0.50000kg
Descrizione
For use with:- ceramus™
- solarus™
- opus™
- ceramus™ classic for 30 + 60 mL
- EM dispenser for 10-60 mL akku drive™
Specifica
Suction Valve | |
Suction, Ceramus Classic to 10 mL EM-dispenser 2 to 10 mL and Battery-Drive o.R. |