Learn More
Memmert™ IQ/OQ Document with Device-Specific Works Test Data - Temperature and Humidity
For one free-selectable temperature and humidity value, Includes temperature distribution survey at Memmert to DIN 12880:2007-05
Marca: Memmert™ HPP410ecoD00136
Dettagli aggiuntivi : Peso : 0.01100kg
Descrizione
- 27 measuring points (sizes 110 - 1060, 2200)
- 26 measuring points (size 1400)
- PQ checklist as support for validation by customer
Specifica
HPP410eco | |
IQ/OQ Document with Device-specific Works Test Data |