Learn More
Iridium AAS Standard, 1000 mg/L Ir in HCl10%, Honeywell Fluka™
Sfoglia Tutto Honeywell Fluka Prodotti
Click to view available options
Quantità:
100 mL
500 mL
Dimensione della confezione:
100mL
500mL
Descrizione
Honeywell Fluka™ analytical standards offer a growing range of high quality inorganic and organic reference materials for the European market produced according to ISO 9001. The products are suitable for a wide range of industries and applications and come complete with a detailed Certificate of Analysis (CoA) in every delivery, and are eligible for free expert technical support ensuring you have the information you need to quickly start your analysis and pass an audit.
Specifica
Specifica
| Materiale o nome chimico | Iridium AAS Standard |
| Densità | 1.02 g/cm3 |
| pH | <2 |
| Quantità | 500 mL |