missing translation for 'onlineSavingsMsg'
Learn More

Isatin, 98%, Thermo Scientific Chemicals

Catalog Number p-7020490
Click to view available options
Quantità:
100 g
500 g
2500 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
91-56-5
C8H5NO2
147.133
MFCD00005718
JXDYKVIHCLTXOP-UHFFFAOYSA-N
isatin, indoline-2,3-dione, 2,3-indolinedione, indole-2,3-dione, isatine, 2,3-dioxoindoline, pseudoisatin, tribulin, isotin, isatic acid lactam
7054
CHEBI:27539
1H-indole-2,3-dione
C1=CC=C2C(=C1)C(=O)C(=O)N2
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
91-56-5
C8H5NO2
147.133
MFCD00005718
JXDYKVIHCLTXOP-UHFFFAOYSA-N
isatin, indoline-2,3-dione, 2,3-indolinedione, indole-2,3-dione, isatine, 2,3-dioxoindoline, pseudoisatin, tribulin, isotin, isatic acid lactam
7054
CHEBI:27539
1H-indole-2,3-dione
C1=CC=C2C(=C1)C(=O)C(=O)N2

Isatin is an endogenous monoamine oxidize (MAO) inhibitor involved in stress and anxiety. Additionally, isatin inhibits alkaline phosphatase (ALP), nitric oxide (NO)-stimulated soluble guanylate cyclase, and other enzymes. Isatins undergo a one-pot Wolff-Kishner like reduction with hydrazine hydrate, under surprisingly mild conditions, to give the corresponding oxindoles. It is used as chromatographic spray reagent for amino acid detection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isatin is an endogenous monoamine oxidize (MAO) inhibitor involved in stress and anxiety. Additionally, isatin inhibits alkaline phosphatase (ALP), nitric oxide (NO)-stimulated soluble guanylate cyclase, and other enzymes. Isatins undergo a one-pot Wolff-Kishner like reduction with hydrazine hydrate, under surprisingly mild conditions, to give the corresponding oxindoles. It is used as chromatographic spray reagent for amino acid detection.

Solubility
Soluble in water (1.9 g/L at 20°C).

Notes
Store under cool dry place. Ensure good ventilation. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Materiale o nome chimico Isatin
Punto di fusione ∼199°C to 200°C
pH 7
Punto d'infiammabilità 220°C (428°F)
Odore Odorless
Quantità 100 g
Beilstein 383659
Indice di Merck 14,5104
Informazioni di solubilità Soluble in water (1.9g/L at 20°C).
Peso formulazione 147.13
Percent Purity 98%
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'