missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ISL2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 18659-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
ISL2 is a transcriptional factor that defines subclasses of motoneurons that segregate into columns in the spinal cord and select distinct axon pathways.Specifica
ISL2 | |
Polyclonal | |
Unconjugated | |
ISL2 | |
ISL LIM homeobox 2, ISL2, Islet 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
104360, 57233, 64843 | |
-20°C | |
Liquid |
Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P50480, Q96A47, Q9CXV0 | |
Isl2 | |
ISL2 Fusion Protein Ag13333 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |