missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITLN2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA552324
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: TCAFSFSSLP RSCKEIKERC HSAGDGLYFL RTKNGVVY Highest antigen sequence identity to the following orthologs: Mouse - 68%, Rat - 63%.
May play a role in the defense system against pathogens.Specifica
ITLN2 | |
Polyclonal | |
Unconjugated | |
ITLN2 | |
Endothelial lectin HL-2; HL2; HL-2; intelectin 2; intelectin-2; ITLN2; UNQ2789/PRO7179 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
142683 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
Lot-Specific | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8WWU7 | |
ITLN2 | |
Recombinant protein corresponding to Human ITLN2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only