Learn More
BUCHI K-439 Model Speed Digestor
Marca: BUCHI 1154392100
Dettagli aggiuntivi : Peso : 25.37000kg
Specifica
Speed Digestor | |
220/240 V ±10 V | |
620 mm | |
Feed - Kjeldahl, Chemicals/Agricultural, Beverage, Pharmaceutical / Cosmetics , Food , Environmental Analysis | |
310 mm | |
540 mm |
50/60 Hz | |
2000 W | |
220/240 V ±10 V, 50/60 Hz, 1280 W | |
300mL Sample tubes and standard suction module | |
1 Ea. |