Learn More
KBTBD11 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA562334
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: YRLLKYDPRR DEWQECPCSS SRERSADMVA LDGFIYRFDL SGSRGEAQAA GPSGVSVSRY HCLAKQWSPC V Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 86%.
KBTBD11 Polyclonal antibody specifically detects KBTBD11 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifica
KBTBD11 | |
Polyclonal | |
Unconjugated | |
KBTBD11 | |
2900016B01Rik; 4930465M17Rik; Chronic myelogenous leukemia-associated protein; CMLAP; Kbtbd11; Kelch domain-containing protein 7B; kelch repeat and BTB (POZ) domain containing 11; kelch repeat and BTB domain containing 11; kelch repeat and BTB domain-containing protein 11; KIAA0711; KLHDC7C; mKIAA0711 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9920 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O94819 | |
KBTBD11 | |
Recombinant protein corresponding to Human KBTBD11 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only