missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KEAP1 Monoclonal Antibody (OTI1A1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Marca: Origene Technologies TA502056S
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
KEAP1 Monoclonal antibody specifically detects KEAP1 in Human samples. It is validated for Flow Cytometry, Western BlotSpecifica
| KEAP1 | |
| Monoclonal | |
| 0.75 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q13137 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 10241 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Western Blot | |
| OTI1A1 | |
| Unconjugated | |
| CALCOCO2 | |
| cytosolic inhibitor of Nrf2; INrf2; Keap1; kelch like ECH associated protein 1; Kelch-like ECH-associated protein 1; kelch-like family member 19; kelch-like protein 19; Kiaa0132; KLHL19; mKIAA0132; NRF2 cytosolic inhibitor; ring canal protein | |
| Full length human recombinant protein of human CALCOCO2 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction