Learn More
KIAA1984 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 20945-1-AP-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
CCDC183 is a protein coding gene.Specifica
| KIAA1984 | |
| Polyclonal | |
| Unconjugated | |
| CCDC183 | |
| KIAA1984, PARF, RP11 216L13.9 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 84960 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunocytochemistry, Western Blot | |
| 0.26 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q5T5S1 | |
| CCDC183 | |
| KIAA1984 Fusion Protein Ag15053 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |