Learn More
Shield Medicare™ Klerwipe™ Sterile Low Particulate Dry Wipes
Low particulate binder-free hydroentangled polyester/cellulose wipes
Marca: Shield Medicare™ 3035520
Dettagli aggiuntivi : Peso : 1.13740kg
Descrizione
- Each pack of wipes, featuring Klertear, is packed in a Grade C cleanroom before being terminally sterilised by gamma irradiation
Specifica
Low Particulate Dry Wipe | |
400 x 400 mm | |
10 Pk. x 10 Wipes |