missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLF13 Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 18352-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Anti-RANTES factor of Late Activated T Lymphocytes-1 (RFLAT-1) polyclonal antiserum. RFLAT-1 is a novel protein belonging to the TFIIIA-like zinc finger protein superfamily and binds to the A site of the RANTES (Regulated upon Activation, Normal T cell Expressed and Secreted) chemokine. RANTES is involved in the generation of inflammatory infiltrate and HIV entry into immune cells.Specifica
KLF13 | |
Polyclonal | |
Unconjugated | |
KLF13 | |
BTE binding protein 3, BTEB3, FKLF2, KLF13, Krueppel like factor 13, Kruppel like factor 13, NSLP1, RFLAT 1, RFLAT1, Transcription factor BTEB3, Transcription factor NSLP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
499171, 51621 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9Y2Y9 | |
KLF13 | |
KLF13 Fusion Protein Ag13200 | |
20 μL | |
Primary | |
Rat, Human | |
Antibody | |
IgG |