Learn More
Abnova™ KRAS/CEN12q FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
Marca:  Abnova™ FG0017.100uL
Dettagli aggiuntivi : Peso : 0.02000kg
Codice prodotto. 16013005
  • 1475.00€ / 100µL
Spedizione stimata: 21-05-2024
per visualizzare lo stock disponibile
Includes: DAPI Counterstain (1500ng/mL ) 125μL.
Descrizione
Descrizione
FISH is a technique used to identify and localize the presence or absence of specific DNA sequences on cells and tissues. Abnova has developed a range of FISH probes for the detection of gene amplification, loss, and translocation. Each FISH probe has a pair of locus-specific, fluorophore-labeled probes originated from a bacterial chromosome (BAC) library.
Advantages
  • Dual colored probes for fast, sensitive, and specific detection
  • Work on metaphase spread, paraffin embedded, and frozen tissue
  • Identify gene amplification, loss, and translocation
  • High signal to noise ratio
  • Low cross reactivity
Specifica
Specifica
Store at 4°C in the dark.
DAPI Counterstain (1500 ng/mL ) 250 uL
100 μL
Fluorescent In Situ Hybridization (Cell), Fluorescent In Situ Hybridization (Formalin/PFA-fixed paraffin-embedded sections)
KRAS/CEN12q FISH Probe