Learn More
Abnova™ KRAS/CEN12q FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
Marca: Abnova™ FG0017.100uL
Dettagli aggiuntivi : Peso : 0.02000kg
Includes: DAPI Counterstain (1500ng/mL ) 125μL.
Descrizione
FISH is a technique used to identify and localize the presence or absence of specific DNA sequences on cells and tissues. Abnova has developed a range of FISH probes for the detection of gene amplification, loss, and translocation. Each FISH probe has a pair of locus-specific, fluorophore-labeled probes originated from a bacterial chromosome (BAC) library.
Advantages
- Dual colored probes for fast, sensitive, and specific detection
- Work on metaphase spread, paraffin embedded, and frozen tissue
- Identify gene amplification, loss, and translocation
- High signal to noise ratio
- Low cross reactivity
Specifica
Store at 4°C in the dark. | |
DAPI Counterstain (1500 ng/mL ) 250 uL | |
100 μL |
Fluorescent In Situ Hybridization (Cell), Fluorescent In Situ Hybridization (Formalin/PFA-fixed paraffin-embedded sections) | |
KRAS/CEN12q FISH Probe |