missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KU70/XRCC6 Mouse anti-Human, Mouse, Rat, Clone: 3C12D9, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66607-1-IG-150UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
XRCC6 is a nuclear complex consisting of two subunits with molecular masses of approximately 70 and 80 kDa. The complex functions as a single-stranded DNA-dependent ATP-dependent helicase. The complex may be involved in the repair of nonhomologous DNA ends such as that required for double-strand break repair, transposition, and V(D)J recombination. High levels of autoantibodies to p70 and p80 have been found in some patients with systemic lupus erythematosus.Specifica
KU70/XRCC6 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P12956, P23475 | |
Xrcc6 | |
KU70,XRCC6 Fusion Protein Ag21301 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
3C12D9 | |
Unconjugated | |
XRCC6 | |
5 dRP lyase Ku70, 70 kDa subunit of Ku antigen, CTC75, CTCBF, DNA repair protein XRCC6, G22P1, KU70, ML8, Thyroid lupus autoantigen, TLAA, XRCC6 | |
Mouse | |
Protein G | |
RUO | |
14375, 25019, 2547 | |
-20°C | |
Liquid |