Learn More
L 700 Technical Buffer Solution, SI Analytics™
SI Analytics™ L 700 Technical Buffer Solution is supplied in unique double-end ampoules that offer a particularly high degree of reliability and measuring accuracy.
Marca: SI Analytics 285138735
Dettagli aggiuntivi : Peso : 0.25000kg
Descrizione
- Hot steam sterilized for longer stability
- No preservation agents used
- The exactness of the pH measurement is mainly dependent on the accuracy of calibration
- Hermetically sealed in the glass ampoule and sterilized with hot steam
- The ampoules can be easily opened at the breaking point
- Highest measurement reliability
- Extremely long storage times, thanks to hot-steam sterilization
- No preservative agents
- Maximize calibration reliability
Specifica
With manufacturer's certificate | |
1000 mL | |
DURAN™ Glass Bottle |
7 | |
Technical Buffer |