Learn More
L 700 Technical Buffer Solution, SI Analytics™
SI Analytics™ L 700 Technical Buffer Solution is supplied in unique double-end ampoules that offer a particularly high degree of reliability and measuring accuracy.
Marca:  SI Analytics 285138735
Dettagli aggiuntivi : Peso : 0.25000kg
Codice prodotto. 10559561
Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.
Descrizione
Descrizione
  • Hot steam sterilized for longer stability
  • No preservation agents used
  • The exactness of the pH measurement is mainly dependent on the accuracy of calibration
  • Hermetically sealed in the glass ampoule and sterilized with hot steam
  • The ampoules can be easily opened at the breaking point
  • Highest measurement reliability
  • Extremely long storage times, thanks to hot-steam sterilization
  • No preservative agents
  • Maximize calibration reliability
Specifica
Specifica
With manufacturer's certificate
1000 mL
DURAN™ Glass Bottle
7
Technical Buffer